Assay Details
Target Gene Details
| Entrez Gene ID: | 3788 | 
| Gene Name: | potassium voltage-gated channel modifier subfamily S member 2 | 
| Gene Aliases: | KV9.2 | 
| Location: | Chr.8:98427022-98430797 on Build GRCh38 | 
| Assay Gene Location: | Within Exon 2 | 
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID | 
|---|---|---|---|---|
| KCNS2 | NM_020697.3 | 2 | 743 | NP_065748.1 | 
| AB032970.1 | 2 | 548 | ||
| AK293096.1 | 1 | 687 | ||
| BC027932.1 | 2 | 716 | AAH27932.1 | |
| BC034778.1 | 2 | 743 | AAH34778.1 | 
Target Copy Number Variation Details
| DGV Version: | Release date: 2016-05-15, GRCh GRCh38 | 
| Target Variation | Location | CNV Subtype | Genes | 
|---|---|---|---|
| nsv1034492 | Chr.8:98424113 - 98444288 on Build GRCh38 | Gain |  KCNS2  STK3 | 
| nsv831406 | Chr.8:98278517 - 98469848 on Build GRCh38 | Loss |  KCNS2  NIPAL2  STK3 | 
 Back To Top
 Back To TopMore Information
| Set Membership: |  Intragenic  Exonic  DGV Variation | 
Panther Classification:
Gene Ontology Categories:
 Back To Top
 Back To Top
 Genomic Map
Genomic Map Back To Top
 Back To Top