Assay Details
Target Gene Details
Entrez Gene ID: | 3932 |
Gene Name: | LCK proto-oncogene, Src family tyrosine kinase |
Gene Aliases: |
IMD22, LSK, YT16, p56lck, pp58lck |
Location: |
Chr.1:32251239-32286167 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LCK | NM_005356.4 | 1 | 84 | NP_005347.3 |
| XM_011541453.2 | 1 | 84 | XP_011539755.1 | |
| XM_011541455.2 | 1 | 84 | XP_011539757.1 | |
| AK098027.1 | ||||
| DA007906.1 | 1 | 84 | ||
| M36881.1 | AAA59502.1 | |||
| X05027.1 | CAA28691.1 | |||
| X13529.1 | CAA31884.1 |
More Information
Set Membership: |
|

Genomic Map