Assay Details
Target Gene Details
Entrez Gene ID: | 4058 |
Gene Name: | leukocyte receptor tyrosine kinase |
Gene Aliases: |
TYK1 |
Location: |
Chr.15:41503639-41513887 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LTK | NM_001135685.1 | NP_001129157.1 | ||
| NM_002344.5 | 11 | 1609 | NP_002335.2 | |
| NM_206961.3 | 10 | 1426 | NP_996844.1 | |
| XM_011521556.2 | 11 | 1657 | XP_011519858.1 | |
| XM_011521557.2 | 10 | 1489 | XP_011519859.1 | |
| XM_017022181.1 | 11 | 1672 | XP_016877670.1 | |
| XM_017022182.1 | 10 | 1528 | XP_016877671.1 | |
| XM_017022183.1 | XP_016877672.1 | |||
| AK296892.1 | ||||
| AK314006.1 | 11 | 1576 | ||
| BC045607.1 | 10 | 1853 | AAH45607.1 | |
| D16105.1 | 11 | 1528 | BAA03679.1 | |
| X52213.1 | 4 | 492 | CAA36460.1 | |
| X60702.1 | 10 | 1418 | CAA43113.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv471241 | Chr.15:41491616 - 41528002 on Build GRCh38 | Loss |
|
| esv33378 | Chr.15:41494307 - 41513562 on Build GRCh38 | Gain |
|
| nsv524476 | Chr.15:41397034 - 41528002 on Build GRCh38 | Loss |
|
| nsv569233 | Chr.15:41492611 - 41513039 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map