Assay Details
Target Gene Details
Entrez Gene ID: | 257068 |
Gene Name: | phosphatidylinositol specific phospholipase C X domain containing 2 |
Gene Aliases: |
- |
Location: |
Chr.3:111674676-111846447 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PLCXD2 | NM_001185106.1 | 1 | 123 | NP_001172035.1 |
| NM_153268.3 | 1 | 123 | NP_695000.1 | |
| AK056141.1 | 1 | 123 | BAB71103.1 | |
| AL712245.1 | 1 | 140 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv591275 | Chr.3:110918624 - 111782250 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map