Assay Details
Target Gene Details
Entrez Gene ID: | 2067 |
Gene Name: | ERCC excision repair 1, endonuclease non-catalytic subunit |
Gene Aliases: |
COFS4, RAD10, UV20 |
Location: |
Chr.19:45407333-45478866 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ERCC1 | XM_005258636.4 | XP_005258693.1 | ||
| XM_017026464.1 | XP_016881953.1 | |||
| AK092039.1 |
Target Gene Details
Entrez Gene ID: | 2354 |
Gene Name: | FosB proto-oncogene, AP-1 transcription factor subunit |
Gene Aliases: |
AP-1, G0S3, GOS3, GOSB |
Location: |
Chr.19:45467995-45475179 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FOSB | NM_001114171.1 | 2 | 778 | NP_001107643.1 |
| NM_006732.2 | 2 | 778 | NP_006723.2 | |
| XM_005258691.1 | 2 | 777 | XP_005258748.1 | |
| AK225070.1 | 2 | 778 | ||
| AK292720.1 | 3 | 377 | ||
| BC036724.1 | 3 | 757 | AAH36724.1 | |
| BC040197.1 | 1 | 781 | AAH40197.1 | |
| BX538117.1 | 2 | 1062 | ||
| CR542101.1 | 3 | 186 | CAG46898.1 | |
| EU178109.1 | 3 | 779 | ||
| EU178110.1 | 3 | 779 | ||
| EU178111.1 | ||||
| EU178112.1 | ||||
| EU178113.1 | 3 | 779 | ||
| EU178114.1 | ||||
| EU178115.1 | ||||
| EU178116.1 | ||||
| L49169.1 | 3 | 779 | AAB53946.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833848 | Chr.19:45457372 - 45644939 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map