Assay Details
Target Gene Details
Entrez Gene ID: | 419 |
Gene Name: | ADP-ribosyltransferase 3 |
Gene Aliases: |
ARTC3 |
Location: |
Chr.4:76011180-76112802 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ART3 | NM_001130017.2 | NP_001123489.1 | ||
| AK225860.1 | ||||
| AY543656.1 | AAT10278.1 | |||
| AY543657.1 | AAT10279.1 | |||
| AY543658.1 | AAT10280.1 | |||
| AY543659.1 | AAT10281.1 | |||
| DB451386.1 | ||||
| U47054.1 | AAB01894.1 |
Target Gene Details
Entrez Gene ID: | 3627 |
Gene Name: | C-X-C motif chemokine ligand 10 |
Gene Aliases: |
C7, IFI10, INP10, IP-10, SCYB10, crg-2, gIP-10, mob-1 |
Location: |
Chr.4:76021116-76023536 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CXCL10 | NM_001565.3 | NP_001556.2 | ||
| AA321649.1 | ||||
| BC010954.1 | AAH10954.1 | |||
| X02530.1 | CAA26370.1 |
More Information
Set Membership: |
|

Genomic Map