Assay Details
Target Gene Details
Entrez Gene ID: | 4593 |
Gene Name: | muscle associated receptor tyrosine kinase |
Gene Aliases: |
CMS9, FADS |
Location: |
Chr.9:110668189-110806633 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MUSK | NM_001166280.1 | 2 | 276 | NP_001159752.1 |
| NM_001166281.1 | 2 | 276 | NP_001159753.1 | |
| NM_005592.3 | 2 | 276 | NP_005583.1 | |
| XM_005251994.3 | 2 | 858 | XP_005252051.1 | |
| XM_005251995.3 | 2 | 857 | XP_005252052.1 | |
| XM_005251996.3 | 2 | 856 | XP_005252053.1 | |
| XM_017014734.1 | 2 | 858 | XP_016870223.1 | |
| AA448972.1 | 2 | 241 | ||
| AF006464.1 | 2 | 188 | AAB63044.1 | |
| BC109098.1 | 2 | 188 | AAI09099.1 | |
| BC109099.1 | 2 | 188 | AAI09100.1 | |
| BG825304.1 | 2 | 277 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv466482 | Chr.9:110379729 - 110713825 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map