Assay Details
Target Gene Details
Entrez Gene ID: | 4323 |
Gene Name: | matrix metallopeptidase 14 |
Gene Aliases: |
MMP-14, MMP-X1, MT-MMP, MT-MMP 1, MT1-MMP, MT1MMP, MTMMP1, WNCHRS |
Location: |
Chr.14:22836533-22847600 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MMP14 | NM_004995.3 | 10 | 1952 | NP_004986.1 |
| AK291325.1 | 10 | 1894 | ||
| AK312657.1 | 10 | 1900 | ||
| BC064803.1 | 10 | 1886 | AAH64803.1 | |
| D26512.1 | 10 | 1778 | BAA05519.1 | |
| HE978355.1 | 10 | 1901 | ||
| U41078.1 | 10 | 1667 | AAA83770.1 | |
| X83535.1 | 10 | 1786 | CAA58519.1 | |
| X90925.1 | 10 | 1779 | CAA62432.1 | |
| Z48481.1 | 10 | 1780 | CAA88372.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv563998 | Chr.14:22414183 - 22879743 on Build GRCh38 | Gain |
|
| nsv952816 | Chr.14:22837792 - 22853491 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map