Assay Details
Target Gene Details
Entrez Gene ID: | 3249 |
Gene Name: | hepsin |
Gene Aliases: |
TMPRSS1 |
Location: |
Chr.19:35040506-35066573 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HPN | NM_002151.2 | 6 | 463 | NP_002142.1 |
| NM_182983.2 | 5 | 1043 | NP_892028.1 | |
| XM_005258838.4 | 5 | 541 | XP_005258895.2 | |
| XM_006723181.3 | 5 | 417 | XP_006723244.2 | |
| XM_017026731.1 | 5 | 716 | XP_016882220.1 | |
| XM_017026732.1 | 4 | 278 | XP_016882221.1 | |
| AK095160.1 | 2 | 259 | ||
| AK293622.1 | 4 | 297 | ||
| AK315655.1 | 5 | 396 | ||
| BC025716.1 | 5 | 415 | AAH25716.1 | |
| EU644753.1 | 4 | 218 | ||
| M18930.1 | 6 | 463 | AAA36013.1 | |
| X07732.1 | 5 | 1043 | CAA30558.1 |
Target Gene Details
Entrez Gene ID: | 100128675 |
Gene Name: | HPN antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.19:35059059-35106304 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HPN-AS1 | NR_024562.1 | 4 | 453 | |
| AK295317.1 | 5 | 453 |
More Information
Set Membership: |
|

Genomic Map