Assay Details
Target Gene Details
Entrez Gene ID: | 58529 |
Gene Name: | myozenin 1 |
Gene Aliases: |
CS-2, FATZ, MYOZ |
Location: |
Chr.10:73631612-73641757 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MYOZ1 | NM_021245.3 | NP_067068.1 | ||
| AF240633.1 | AAG24509.1 | |||
| AJ278124.1 | CAB92967.1 | |||
| AK300683.1 | ||||
| AK308200.1 | ||||
| AL359210.1 | CAB94568.1 | |||
| AY013297.1 | AAG38940.1 | |||
| BC025753.1 | AAH25753.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv7143 | Chr.10:73605272 - 73640261 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map