Assay Details
Target Gene Details
Entrez Gene ID: | 375190 |
Gene Name: | family with sequence similarity 228 member B |
Gene Aliases: |
- |
Location: |
Chr.2:24076874-24169638 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM228B | NM_001291328.1 | NP_001278257.1 | ||
| NR_111929.1 | ||||
| BM722814.1 |
Target Gene Details
Entrez Gene ID: | 9540 |
Gene Name: | tumor protein p53 inducible protein 3 |
Gene Aliases: |
PIG3 |
Location: |
Chr.2:24077428-24085215 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TP53I3 | NM_001206802.2 | 4 | 852 | NP_001193731.1 |
| NM_004881.4 | 5 | 1933 | NP_004872.2 | |
| NM_147184.3 | 6 | 1534 | NP_671713.1 | |
| XM_005264650.2 | 6 | 1406 | XP_005264707.1 | |
| XM_006712150.2 | 5 | 1337 | XP_006712213.1 | |
| AF010309.1 | 5 | 1571 | AAC39528.1 | |
| AK223382.1 | 5 | 1770 | BAD97102.1 | |
| AK297579.1 | 5 | 1374 | ||
| BC000474.2 | 6 | 1104 | AAH00474.1 | |
| BT007149.1 | AAP35813.1 | |||
| CN285566.1 | 2 | 375 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv997445 | Chr.2:23357900 - 24374347 on Build GRCh38 | Gain |
|
| nsv997725 | Chr.2:24052436 - 25123406 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map