Assay Details
Target Gene Details
Entrez Gene ID: | 9329 |
Gene Name: | general transcription factor IIIC subunit 4 |
Gene Aliases: |
KAT12, TF3C-delta, TFIII90, TFIIIC290, TFIIIC90, TFIIICDELTA |
Location: |
Chr.9:132670035-132694955 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GTF3C4 | NM_012204.3 | 2 | 2609 | NP_036336.2 |
| NR_133925.1 | 2 | 2609 | ||
| XM_017015317.1 | 2 | 2156 | XP_016870806.1 | |
| XM_017015318.1 | XP_016870807.1 | |||
| AF142328.1 | 2 | 2297 | AAF05087.1 | |
| AK024655.1 | 1 | 1072 | ||
| BC094774.1 | 2 | 2154 | AAH94774.1 | |
| BC104755.1 | 2 | 2114 | AAI04756.1 | |
| BC112245.1 | 2 | 2114 | AAI12246.1 |
More Information
Set Membership: |
|

Genomic Map