Assay Details
Target Gene Details
Entrez Gene ID: | 2870 |
Gene Name: | G protein-coupled receptor kinase 6 |
Gene Aliases: |
GPRK6 |
Location: |
Chr.5:177426496-177442849 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 5 - Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GRK6 | NM_001004105.2 | NP_001004105.1 | ||
| NM_001004106.2 | NP_001004106.1 | |||
| NM_002082.3 | NP_002073.2 | |||
| XM_006714858.3 | XP_006714921.1 | |||
| XM_006714859.2 | XP_006714922.1 | |||
| XM_011534537.2 | XP_011532839.1 | |||
| AF040751.1 | AAC09273.1 | |||
| AF040752.1 | AAC09274.1 | |||
| AK056697.1 | ||||
| BC009277.2 | AAH09277.1 | |||
| BC017272.1 | AAH17272.1 | |||
| BI258860.1 | ||||
| BU507609.1 | ||||
| BX382886.2 | ||||
| L16862.1 | AAA60175.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv471057 | Chr.5:177310308 - 177469411 on Build GRCh38 | Loss |
|
| nsv600364 | Chr.5:177299634 - 177432847 on Build GRCh38 | Loss |
|
| nsv1161313 | Chr.5:177356552 - 177463415 on Build GRCh38 | Duplication |
|
| dgv733n27 | Chr.5:177357511 - 177469411 on Build GRCh38 | Loss |
|
| nsv1161311 | Chr.5:177305104 - 177546474 on Build GRCh38 | Duplication |
|
More Information
Set Membership: |
|

Genomic Map