Assay Details
Target Gene Details
Entrez Gene ID: | 60401 |
Gene Name: | ectodysplasin A2 receptor |
Gene Aliases: |
EDA-A2R, EDAA2R, TNFRSF27, XEDAR |
Location: |
Chr.X:66594384-66639303 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EDA2R | NM_001199687.2 | 7 | 2716 | NP_001186616.1 |
| NM_001242310.1 | 7 | 2681 | NP_001229239.1 | |
| NM_001324199.1 | 5 | 2362 | NP_001311128.1 | |
| NM_001324201.1 | 6 | 2636 | NP_001311130.1 | |
| NM_001324202.1 | 5 | 2065 | NP_001311131.1 | |
| NM_001324204.1 | 5 | 2164 | NP_001311133.1 | |
| NM_001324205.1 | 5 | 2158 | NP_001311134.1 | |
| NM_001324206.1 | 7 | 2724 | NP_001311135.1 | |
| NM_021783.4 | 7 | 2718 | NP_068555.1 | |
| NR_136726.1 | 7 | 2625 | ||
| NR_136727.1 | 7 | 2627 | ||
| XM_011531000.1 | 8 | 2731 | XP_011529302.1 | |
| XM_011531001.1 | 8 | 2779 | XP_011529303.1 | |
| XM_011531002.1 | 8 | 2436 | XP_011529304.1 | |
| XM_011531003.2 | 6 | 2531 | XP_011529305.1 | |
| XM_011531005.2 | 5 | 2365 | XP_011529307.1 | |
| XM_011531006.2 | 6 | 2230 | XP_011529308.1 | |
| XM_017029703.1 | 6 | 2628 | XP_016885192.1 | |
| XM_017029704.1 | 7 | 2373 | XP_016885193.1 | |
| XM_017029705.1 | 6 | 2506 | XP_016885194.1 | |
| XM_017029706.1 | 6 | 2281 | XP_016885195.1 | |
| XM_017029707.1 | 6 | 2415 | XP_016885196.1 | |
| XM_017029708.1 | 5 | 2057 | XP_016885197.1 | |
| BC034919.1 | 7 | 2665 | AAH34919.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv2305e212 | Chr.X:66550620 - 66717352 on Build GRCh38 | Gain |
|
| nsv520630 | Chr.X:66486237 - 66797185 on Build GRCh38 | Gain+Loss |
|
| esv3576919 | Chr.X:65837678 - 66804698 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map