Assay Details
Target Gene Details
Entrez Gene ID: | 90427 |
Gene Name: | Bcl2 modifying factor |
Gene Aliases: |
- |
Location: |
Chr.15:40087890-40108884 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BMF | NM_001003940.1 | 5 | 2913 | NP_001003940.1 |
| NM_001003942.1 | 3 | 2615 | NP_001003942.1 | |
| NM_001003943.2 | 4 | 2762 | NP_001003943.1 | |
| NM_033503.3 | 4 | 2764 | NP_277038.1 | |
| AK024472.1 | 4 | 2774 | ||
| BC060783.1 | 5 | 2911 | AAH60783.1 | |
| BC070043.1 | 5 | 2911 | ||
| BQ064116.1 | 1 | 132 |
More Information
Set Membership: |
|

Genomic Map