Assay Details
Target Gene Details
Entrez Gene ID: | 64978 |
Gene Name: | mitochondrial ribosomal protein L38 |
Gene Aliases: |
HSPC262, L38MT, MRP-L3, MRP-L38, RPML3 |
Location: |
Chr.17:75898643-75905100 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MRPL38 | NM_032478.3 | NP_115867.2 | ||
| AF161380.1 | AAF28940.1 | |||
| AK313283.1 | ||||
| BC013311.2 | AAH13311.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833545 | Chr.17:75738140 - 75945931 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map