Assay Details
Target Gene Details
Entrez Gene ID: | 92935 |
Gene Name: | methionyl-tRNA synthetase 2, mitochondrial |
Gene Aliases: |
COXPD25, MetRS, mtMetRS |
Location: |
Chr.2:197705304-197708390 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MARS2 | NM_138395.3 | 1 | 1651 | NP_612404.1 |
| AB107013.1 | 1 | 1594 | BAC92749.1 | |
| AK098121.1 | 1 | 1586 | BAC05238.1 | |
| AK301252.1 | 2 | 1366 | ||
| BC009115.1 | 1 | 1554 | AAH09115.1 | |
| BC040934.2 | 1 | 629 | AAH40934.1 | |
| BC126294.1 | 1 | 1589 |
More Information
Set Membership: |
|

Genomic Map