Assay Details
Target Gene Details
Entrez Gene ID: | 64376 |
Gene Name: | IKAROS family zinc finger 5 |
Gene Aliases: |
PEGASUS, ZNFN1A5 |
Location: |
Chr.10:122990806-123008850 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IKZF5 | NM_001271840.1 | 3 | 375 | NP_001258769.1 |
| NR_073475.1 | 3 | 419 | ||
| NR_073476.1 | 3 | 419 | ||
| NR_073477.1 | 3 | 406 | ||
| NR_073478.1 | 2 | 268 | ||
| NR_073479.1 | 4 | 527 | ||
| XM_006717947.3 | 4 | 489 | XP_006718010.1 | |
| XM_017016550.1 | 3 | 367 | XP_016872039.1 | |
| XM_017016551.1 | 4 | 463 | XP_016872040.1 | |
| XM_017016552.1 | 3 | 355 | XP_016872041.1 | |
| XM_017016553.1 | 3 | 393 | XP_016872042.1 | |
| XM_017016554.1 | 4 | 501 | XP_016872043.1 | |
| XM_017016555.1 | 3 | 380 | XP_016872044.1 | |
| AF230808.1 | 1 | 65 | AAG39220.1 | |
| AK026626.1 | 3 | 364 | BAB15512.1 | |
| AK122899.1 | 4 | 2166 | ||
| AK226143.1 | 3 | 360 | ||
| BX647958.1 | 3 | 368 | CAH56199.1 | |
| CR749800.1 | 3 | 361 | CAH18660.1 | |
| DA218215.1 | 3 | 339 | ||
| DA227834.1 | 2 | 223 | ||
| DB080324.1 | 3 | 406 | ||
| DB224006.1 | 4 | 472 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv7586 | Chr.10:122982687 - 123016052 on Build GRCh38 | Insertion |
|
| nsv832010 | Chr.10:122826964 - 123037874 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map