Assay Details
Target Gene Details
Entrez Gene ID: | 64135 |
Gene Name: | interferon induced with helicase C domain 1 |
Gene Aliases: |
AGS7, Hlcd, IDDM19, MDA-5, MDA5, RLR-2, SGMRT1 |
Location: |
Chr.2:162267079-162318708 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 14 - Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IFIH1 | NM_022168.3 | NP_071451.2 | ||
| AF095844.1 | AAG34368.1 | |||
| AK056293.1 | BAB71141.1 | |||
| AK292941.1 | ||||
| AK314611.1 | ||||
| AY017378.1 | AAG54076.1 | |||
| BC111750.1 | AAI11751.1 | |||
| BQ772836.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv583520 | Chr.2:162267541 - 162611723 on Build GRCh38 | Loss |
|
| nsv834440 | Chr.2:162145758 - 162318828 on Build GRCh38 | Gain |
|
| esv3593177 | Chr.2:162204243 - 162287181 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map