Assay Details
Target Gene Details
Entrez Gene ID: | 64115 |
Gene Name: | chromosome 10 open reading frame 54 |
Gene Aliases: |
B7-H5, B7H5, DD1alpha, GI24, PD-1H, PP2135, SISP1, VISTA |
Location: |
Chr.10:71747556-71773580 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C10orf54 | NM_022153.1 | 6 | 951 | NP_071436.1 |
| AF193048.1 | 1 | 90 | AAG22476.1 | |
| AK024449.1 | 6 | 869 | ||
| AY265805.1 | 6 | 866 | AAP03084.1 | |
| AY358379.1 | 6 | 951 | AAQ88745.1 | |
| BC020568.1 | 6 | 823 | AAH20568.1 | |
| BC111048.1 | 6 | 799 | AAI11049.1 | |
| BC127257.1 | 3 | 229 | ||
| EF506490.1 | 6 | 863 |
Target Gene Details
Entrez Gene ID: | 64072 |
Gene Name: | cadherin-related 23 |
Gene Aliases: |
CDHR23, USH1D |
Location: |
Chr.10:71396934-71815947 on Build GRCh38 |
Assay Gene Location: | Within Intron 46 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831910 | Chr.10:71635558 - 71843223 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map