Assay Details
Target Gene Details
Entrez Gene ID: | 105373764 |
Gene Name: | uncharacterized LOC105373764 |
Gene Aliases: |
- |
Location: |
Chr.2:177673322-177723289 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC105373764 | NR_136171.1 | |||
| BX094187.1 |
Target Gene Details
Entrez Gene ID: | 50940 |
Gene Name: | phosphodiesterase 11A |
Gene Aliases: |
PPNAD2 |
Location: |
Chr.2:177623249-178108339 on Build GRCh38 |
Assay Gene Location: | Within Exon 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PDE11A | NM_001077196.1 | 12 | 1115 | NP_001070664.1 |
| NM_001077197.1 | 16 | 1645 | NP_001070665.1 | |
| NM_001077358.1 | 14 | 1355 | NP_001070826.1 | |
| NM_016953.3 | 15 | 2614 | NP_058649.3 | |
| AB036704.1 | 15 | 2614 | BAB16371.1 | |
| AB038041.1 | 16 | 1645 | BAB16372.1 | |
| AF281865.1 | 14 | 1327 | AAG32023.1 | |
| AJ251509.1 | 12 | 1114 | CAB82573.1 | |
| AJ278682.1 | 16 | 1688 | CAC15567.1 | |
| AK074273.1 | 5 | 511 | ||
| BC112393.1 | 15 | 2605 | ||
| BC114431.1 | 15 | 2605 | ||
| BX649056.1 | 14 | 1339 | ||
| HM771389.1 | 15 | 2296 | ||
| HM771390.1 | 15 | 2296 | ||
| HM771391.1 | 15 | 2296 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv4109n100 | Chr.2:177671194 - 177721880 on Build GRCh38 | Gain |
|
| esv2762967 | Chr.2:177682191 - 177712274 on Build GRCh38 | Gain |
|
| esv3593466 | Chr.2:177618127 - 177839958 on Build GRCh38 | Gain |
|
| dgv4110n100 | Chr.2:177685321 - 177712274 on Build GRCh38 | Gain |
|
| esv3575314 | Chr.2:177685413 - 177720934 on Build GRCh38 | Gain |
|
| nsv528219 | Chr.2:177687196 - 177702423 on Build GRCh38 | Gain |
|
| esv3593468 | Chr.2:177684859 - 177715374 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map