Assay Details
Target Gene Details
Entrez Gene ID: | 8659 |
Gene Name: | aldehyde dehydrogenase 4 family member A1 |
Gene Aliases: |
ALDH4, P5CD, P5CDh |
Location: |
Chr.1:18871430-18902799 on Build GRCh38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ALDH4A1 | NM_001161504.1 | 15 | 2355 | NP_001154976.1 |
| NM_001319218.1 | 14 | 2342 | NP_001306147.1 | |
| NM_003748.3 | 15 | 2495 | NP_003739.2 | |
| NM_170726.2 | NP_733844.1 | |||
| AK222486.1 | 15 | 2251 | BAD96206.1 | |
| BC007581.1 | 15 | 2225 | AAH07581.1 | |
| BC023600.2 | 15 | 2236 | AAH23600.1 | |
| BX649081.1 | 2 | 1600 | ||
| FJ462711.1 | 14 | 2080 | ||
| U24266.1 | 15 | 2249 | AAC50500.1 | |
| U24267.1 | AAC50501.1 |
More Information
Set Membership: |
|

Genomic Map