Assay Details
Target Gene Details
Entrez Gene ID: | 51304 |
Gene Name: | zinc finger DHHC-type containing 3 |
Gene Aliases: |
DHHC-3, GODZ, ZNF373 |
Location: |
Chr.3:44915261-44976190 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834675 | Chr.3:44805220 - 44966381 on Build GRCh38 | Gain |
|
| nsv834676 | Chr.3:44928707 - 45149356 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map