Assay Details
Target Gene Details
Entrez Gene ID: | 23428 |
Gene Name: | solute carrier family 7 member 8 |
Gene Aliases: |
LAT2, LPI-PC1 |
Location: |
Chr.14:23125295-23183660 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC7A8 | NM_001267036.1 | 9 | 1971 | NP_001253965.1 |
| NM_001267037.1 | 8 | 1716 | NP_001253966.1 | |
| NM_012244.3 | 11 | 2932 | NP_036376.2 | |
| NM_182728.2 | 9 | 1870 | NP_877392.1 | |
| NR_049767.1 | 7 | 1911 | ||
| AB037669.1 | 11 | 2895 | BAB21519.1 | |
| AF135830.1 | 7 | 1513 | AAF05697.1 | |
| AF171669.1 | 11 | 2409 | AAF20381.1 | |
| AK094550.1 | 3 | 1417 | ||
| AL365342.1 | 6 | 2264 | ||
| AL365344.1 | 5 | 1247 | ||
| AL365345.1 | 7 | 1549 | ||
| AL365346.1 | 3 | 1135 | ||
| AL365347.1 | 2 | 523 | ||
| BC036825.1 | 7 | 1762 | AAH36825.1 | |
| BC052250.1 | 11 | 2215 | AAH52250.1 | |
| Y18483.1 | 11 | 2916 | CAB40137.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv564040 | Chr.14:23126512 - 23163297 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map