Assay Details
Target Gene Details
Entrez Gene ID: | 4329 |
Gene Name: | aldehyde dehydrogenase 6 family member A1 |
Gene Aliases: |
MMSADHA, MMSDH |
Location: |
Chr.14:74057662-74084493 on Build GRCh38 |
Assay Gene Location: | Within Intron 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ALDH6A1 | NM_001278593.1 | NP_001265522.1 | ||
| NM_001278594.1 | NP_001265523.1 | |||
| NM_005589.3 | NP_005580.1 | |||
| XM_017021331.1 | XP_016876820.1 | |||
| AF148505.1 | AAF04489.1 | |||
| AF159889.1 | AAF80380.1 | |||
| AJ249994.1 | CAB76468.1 | |||
| AK222687.1 | BAD96407.1 | |||
| AK223244.1 | BAD96964.1 | |||
| AK294243.1 | ||||
| AK311478.1 | ||||
| AK312389.1 | ||||
| BC004909.1 | AAH04909.1 | |||
| BC032371.1 | AAH32371.1 | |||
| M93405.1 | AAA36328.1 |
Target Gene Details
Entrez Gene ID: | 80127 |
Gene Name: | basal body orientation factor 1 |
Gene Aliases: |
C14orf45, CCDC176 |
Location: |
Chr.14:74019350-74078351 on Build GRCh38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BBOF1 | NM_025057.2 | 12 | 2009 | NP_079333.2 |
| XM_011537171.2 | 13 | 2130 | XP_011535473.1 | |
| XM_011537172.1 | XP_011535474.1 | |||
| XM_011537174.2 | 13 | 2123 | XP_011535476.1 | |
| XM_017021659.1 | XP_016877148.1 | |||
| XM_017021660.1 | 11 | 1738 | XP_016877149.1 | |
| XM_017021661.1 | 11 | 1783 | XP_016877150.1 | |
| XM_017021662.1 | 10 | 1567 | XP_016877151.1 | |
| XM_017021663.1 | 10 | 1376 | XP_016877152.1 | |
| AK026842.1 | 5 | 1042 | BAB15572.1 | |
| AK097531.1 | ||||
| AL834487.1 | 7 | 1284 | CAD39145.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv1936n100 | Chr.14:73950133 - 74073431 on Build GRCh38 | Gain |
|
| dgv1938n100 | Chr.14:73984364 - 74075786 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map