Assay Details
Target Gene Details
Entrez Gene ID: | 93436 |
Gene Name: | armadillo repeat containing 6 |
Gene Aliases: |
R30923_1 |
Location: |
Chr.19:19033578-19058178 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ARMC6 | NM_001199196.1 | 1 | 197 | NP_001186125.1 |
| NM_033415.3 | 1 | 197 | NP_219483.1 | |
| XM_005260157.3 | 1 | 165 | XP_005260214.1 | |
| XM_017027484.1 | 1 | 172 | XP_016882973.1 | |
| XM_017027485.1 | 1 | 165 | XP_016882974.1 | |
| AK303914.1 | 1 | 135 | ||
| BC067125.1 | AAH67125.1 | |||
| BX648486.1 | 1 | 92 | ||
| CX869901.1 | 1 | 197 |
Target Gene Details
Entrez Gene ID: | 10147 |
Gene Name: | SURP and G-patch domain containing 2 |
Gene Aliases: |
SFRS14 |
Location: |
Chr.19:18990887-19034052 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SUGP2 | NM_001321697.1 | NP_001308626.1 | ||
| XM_017026137.1 | XP_016881626.1 | |||
| AB002363.2 | ||||
| BC020586.1 | AAH20586.1 | |||
| DA217559.1 | ||||
| DA770920.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv952424 | Chr.19:19033592 - 19069091 on Build GRCh38 | Deletion |
|
| nsv833778 | Chr.19:18901860 - 19090678 on Build GRCh38 | Loss |
|
| nsv1151217 | Chr.19:18627369 - 19284914 on Build GRCh38 | Deletion |
|
| nsv1059305 | Chr.19:19020501 - 19099190 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map