Assay Details
Target Gene Details
Entrez Gene ID: | 204962 |
Gene Name: | solute carrier family 44 member 5 |
Gene Aliases: |
CTL5 |
Location: |
Chr.1:75202131-75724261 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 25 - Exon 26 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC44A5 | NM_001130058.1 | NP_001123530.1 | ||
| NM_001320283.1 | NP_001307212.1 | |||
| NM_001320285.1 | NP_001307214.1 | |||
| NM_001320287.1 | NP_001307216.1 | |||
| NM_152697.4 | NP_689910.2 | |||
| XM_006710445.3 | XP_006710508.1 | |||
| XM_011540986.2 | XP_011539288.1 | |||
| XM_011540987.2 | XP_011539289.1 | |||
| XM_017000607.1 | XP_016856096.1 | |||
| XM_017000608.1 | XP_016856097.1 | |||
| XM_017000609.1 | XP_016856098.1 | |||
| XM_017000610.1 | XP_016856099.1 | |||
| XM_017000611.1 | XP_016856100.1 | |||
| AK091400.1 | BAC03655.1 | |||
| AK093170.1 | BAC04084.1 | |||
| AK128063.1 | ||||
| AK296924.1 | ||||
| AK296941.1 | ||||
| AK299573.1 | ||||
| BC028743.2 | AAH28743.1 | |||
| BC034580.1 | AAH34580.1 | |||
| BC051740.1 | AAH51740.1 |
More Information
Set Membership: |
|

Genomic Map