Assay Details
Target Gene Details
Entrez Gene ID: | 6738 |
Gene Name: | TROVE domain family member 2 |
Gene Aliases: |
RO60, RORNP, SSA2 |
Location: |
Chr.1:193059422-193091777 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TROVE2 | NM_001042369.2 | NP_001035828.1 | ||
| NM_001042370.2 | NP_001035829.2 | |||
| NM_001173524.1 | NP_001166995.1 | |||
| NM_001173525.1 | NP_001166996.1 | |||
| NM_004600.5 | NP_004591.2 | |||
| NR_033393.1 | ||||
| XM_006711495.3 | XP_006711558.1 | |||
| XM_006711496.3 | XP_006711559.1 | |||
| XM_006711497.3 | XP_006711560.1 | |||
| XM_011509922.2 | XP_011508224.1 | |||
| XM_017002180.1 | XP_016857669.1 | |||
| XM_017002181.1 | XP_016857670.1 | |||
| XM_017002182.1 | XP_016857671.1 | |||
| XM_017002183.1 | XP_016857672.1 | |||
| AK314594.1 | ||||
| AY205314.1 | AAO47001.1 | |||
| AY205315.1 | AAO47002.1 | |||
| BC036658.2 | AAH36658.1 | |||
| BX394323.2 | ||||
| CR936683.1 | ||||
| J04137.1 | AAA35493.1 | |||
| M25077.1 | AAA35532.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv498n100 | Chr.1:192992568 - 193114915 on Build GRCh38 | Loss |
|
| nsv548658 | Chr.1:192394185 - 193552205 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map