Assay Details
Target Gene Details
Entrez Gene ID: | 6146 |
Gene Name: | ribosomal protein L22 |
Gene Aliases: |
EAP, HBP15, HBP15/L22, L22 |
Location: |
Chr.1:6185020-6199619 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RPL22 | NM_000983.3 | NP_000974.1 | ||
| AK097075.1 | ||||
| AK311749.1 | ||||
| BC035566.1 | AAH35566.1 | |||
| BC058887.1 | AAH58887.1 | |||
| BC066314.1 | AAH66314.1 | |||
| CA336134.1 | ||||
| CR456873.1 | CAG33154.1 | |||
| D17652.1 | BAA04545.1 | |||
| X59357.1 | CAA42007.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831425 | Chr.1:6129291 - 6264418 on Build GRCh38 | Loss |
|
| nsv945755 | Chr.1:6185921 - 6187830 on Build GRCh38 | Duplication |
|
| nsv545236 | Chr.1:6161915 - 6262876 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map