Assay Details
Target Gene Details
Entrez Gene ID: | 81558 |
Gene Name: | family with sequence similarity 117 member A |
Gene Aliases: |
- |
Location: |
Chr.17:49710325-49764156 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM117A | NM_030802.3 | 8 | 1310 | NP_110429.1 |
| XM_017025182.1 | 8 | 2068 | XP_016880671.1 | |
| AF123073.1 | 8 | 1271 | AAK01477.1 | |
| AK302376.1 | 8 | 1124 | ||
| BC037572.1 | 8 | 1322 | AAH37572.1 | |
| BC065199.1 | 8 | 1275 | AAH65199.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv525469 | Chr.17:49704315 - 49720153 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs114334999] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map