Assay Details
Target Gene Details
Entrez Gene ID: | 387522 |
Gene Name: | TMEM189-UBE2V1 readthrough |
Gene Aliases: |
CROC-1B, CROC1B, KUA-UEV |
Location: |
Chr.20:50081124-50153798 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TMEM189-UBE2V1 | NM_199203.2 | NP_954673.1 | ||
| U39361.1 | AAB72016.1 |
Target Gene Details
Entrez Gene ID: | 7335 |
Gene Name: | ubiquitin conjugating enzyme E2 V1 |
Gene Aliases: |
CIR1, CROC-1, CROC1, UBE2V, UEV-1, UEV1, UEV1A |
Location: |
Chr.20:50081124-50115959 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| UBE2V1 | NM_001257393.1 | 1 | 35 | NP_001244322.1 |
| NM_001257394.1 | 1 | 35 | NP_001244323.1 | |
| NM_001282575.2 | 1 | 35 | NP_001269504.1 | |
| NM_021988.5 | 1 | 35 | NP_068823.2 | |
| NM_022442.5 | 1 | 35 | NP_071887.1 | |
| NM_199144.2 | 1 | 35 | NP_954595.1 | |
| AK314754.1 | ||||
| BU540046.1 | 1 | 36 | ||
| CD103154.1 | ||||
| CN346855.1 | ||||
| DA409547.1 | 1 | 35 | ||
| U97280.1 | AAC02755.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1061982 | Chr.20:50111064 - 50483508 on Build GRCh38 | Loss |
|
| esv3646041 | Chr.20:50114564 - 50116213 on Build GRCh38 | Loss |
|
| nsv833999 | Chr.20:50037107 - 50118431 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map