Assay Details
Target Gene Details
Entrez Gene ID: | 164832 |
Gene Name: | LON peptidase N-terminal domain and ring finger 2 |
Gene Aliases: |
RNF192 |
Location: |
Chr.2:100273291-100322733 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LONRF2 | NM_198461.3 | 12 | 11870 | NP_940863.3 |
| AK056373.1 | 1 | 206 | ||
| AK094044.1 | 1 | 949 | ||
| AK123836.1 | 1 | 1652 | ||
| AK124739.1 | 1 | 2793 | ||
| AK128715.1 | 1 | 3529 | ||
| AL157505.1 | 1 | 104 | ||
| DQ020495.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834311 | Chr.2:100200455 - 100373574 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map