Assay Details
Target Gene Details
Entrez Gene ID: | 56999 |
Gene Name: | ADAM metallopeptidase with thrombospondin type 1 motif 9 |
Gene Aliases: |
- |
Location: |
Chr.3:64515654-64688000 on Build GRCh38 |
Assay Gene Location: | Within Exon 37 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ADAMTS9 | NM_001318781.1 | 36 | 5510 | NP_001305710.1 |
| NM_182920.1 | 37 | 5594 | NP_891550.1 | |
| AF488803.1 | 37 | 5594 | AAO15765.1 | |
| AK300357.1 | 16 | 2266 | ||
| AK303343.1 | 18 | 2729 | ||
| BC130578.1 | 37 | 5584 | ||
| BC171764.1 | 36 | 5500 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv289n21 | Chr.3:64370655 - 64583407 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map