Assay Details
Target Gene Details
Entrez Gene ID: | 158067 |
Gene Name: | adenylate kinase 8 |
Gene Aliases: |
AK 8, C9orf98, DDX31 |
Location: |
Chr.9:132725574-132878811 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AK8 | NM_001317958.1 | NP_001304887.1 | ||
| NM_001317959.1 | NP_001304888.1 | |||
| NM_152572.2 | NP_689785.1 | |||
| BC050576.1 | AAH50576.1 |
Target Gene Details
Entrez Gene ID: | 11092 |
Gene Name: | sperm acrosome associated 9 |
Gene Aliases: |
C9orf9, Mast |
Location: |
Chr.9:132878320-132890314 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SPACA9 | NM_001316898.1 | NP_001303827.1 | ||
| NR_133631.1 | ||||
| BC012940.1 | AAH12940.1 | |||
| DA134573.1 | ||||
| DA281620.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2739138 | Chr.9:132727483 - 135521553 on Build GRCh38 | Deletion |
|
| nsv831743 | Chr.9:132820292 - 132982548 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map