Assay Details
Target Gene Details
Entrez Gene ID: | 288 |
Gene Name: | ankyrin 3, node of Ranvier (ankyrin G) |
Gene Aliases: |
ANKYRIN-G, MRT37 |
Location: |
Chr.10:60026298-60733526 on Build GRCh38 |
Assay Gene Location: | Within Exon 52 |
More Information
Set Membership: |
|

Genomic Map