Assay Details
Target Gene Details
Entrez Gene ID: | 29108 |
Gene Name: | PYD and CARD domain containing |
Gene Aliases: |
ASC, CARD5, TMS, TMS-1, TMS1 |
Location: |
Chr.16:31201486-31202776 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PYCARD | NM_013258.4 | 1 | 273 | NP_037390.2 |
| NM_145182.2 | 1 | 273 | NP_660183.1 | |
| AB023416.2 | 1 | 273 | BAA87339.2 | |
| AF184073.1 | 1 | 261 | AAG01188.1 | |
| AF255794.1 | 1 | 261 | AAF99665.1 | |
| AF310103.1 | 1 | 187 | AAG30286.1 | |
| AF384665.1 | 1 | 240 | AAK63850.1 | |
| AK000211.1 | 1 | 227 | BAA91012.1 | |
| AK314422.1 | 1 | 265 | ||
| BC004470.2 | AAH04470.1 | |||
| BC013569.1 | 1 | 180 | AAH13569.2 |
Target Gene Details
Entrez Gene ID: | 100652740 |
Gene Name: | PYCARD antisense RNA 1 |
Gene Aliases: |
C16orf98, PYCARDOS |
Location: |
Chr.16:31201885-31203452 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PYCARD-AS1 | NR_102400.1 | 2 | 147 | |
| BX464768.2 | 2 | 147 |
More Information
Set Membership: |
|

Genomic Map