Assay Details
Target Gene Details
Entrez Gene ID: | 127943 |
Gene Name: | Fc receptor like B |
Gene Aliases: |
FCRL2, FCRLM2, FCRLY, FREB-2, FcRY |
Location: |
Chr.1:161721544-161728143 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 7 - Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FCRLB | NM_001002901.3 | NP_001002901.1 | ||
| NM_001288829.1 | NP_001275758.1 | |||
| NM_001288830.1 | NP_001275759.1 | |||
| NM_001288831.1 | NP_001275760.1 | |||
| NM_001288832.1 | NP_001275761.1 | |||
| NM_001320241.1 | NP_001307170.1 | |||
| AK055614.1 | 1 | 183 | BAB70971.1 | |
| AK290733.1 | ||||
| AY670683.1 | AAT77987.1 | |||
| AY670684.1 | AAT77988.1 | |||
| AY670685.1 | AAT77989.1 | |||
| AY670686.1 | AAT77990.1 | |||
| AY670687.1 | AAT77991.1 | |||
| AY786314.1 | AAX11390.1 | |||
| BC038564.2 | AAH38564.2 | |||
| BX364259.2 | ||||
| DQ021957.1 | AAY42534.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv428257 | Chr.1:161436525 - 161898473 on Build GRCh38 | Loss |
|
| nsv464184 | Chr.1:161677017 - 161915562 on Build GRCh38 | Gain |
|
| esv2718895 | Chr.1:161723064 - 161726071 on Build GRCh38 | Deletion |
|
| nsv516773 | Chr.1:161721643 - 161732973 on Build GRCh38 | Gain |
|
| esv33393 | Chr.1:161508891 - 162110537 on Build GRCh38 | Gain+Loss |
|
| nsv464195 | Chr.1:161723213 - 161733002 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map