Assay Details
Target Gene Details
Entrez Gene ID: | 84167 |
Gene Name: | chromosome 19 open reading frame 44 |
Gene Aliases: |
- |
Location: |
Chr.19:16496311-16521369 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C19orf44 | NM_001288834.1 | 3 | 1040 | NP_001275763.1 |
| NM_032207.3 | 3 | 1040 | NP_115583.1 | |
| XM_006722920.3 | 3 | 1040 | XP_006722983.1 | |
| XM_011528354.2 | 3 | 1040 | XP_011526656.1 | |
| XM_017027350.1 | 3 | 1040 | XP_016882839.1 | |
| AK025395.1 | 3 | 938 | BAB15125.1 | |
| AK225118.1 | 3 | 957 | ||
| AK225678.1 | 3 | 950 | ||
| AK297781.1 | ||||
| AK311324.1 | ||||
| BC027869.1 | 3 | 912 | AAH27869.1 |
More Information
Set Membership: |
|

Genomic Map