Assay Details
Target Gene Details
Entrez Gene ID: | 56919 |
Gene Name: | DEAH-box helicase 33 |
Gene Aliases: |
DDX33 |
Location: |
Chr.17:5440912-5469060 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 10 - Intron 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DHX33 | NM_001199699.1 | NP_001186628.1 | ||
| NM_020162.3 | NP_064547.2 | |||
| XM_017024877.1 | XP_016880366.1 | |||
| AK025625.1 | BAB15193.1 | |||
| AK026944.1 | BAB15596.1 | |||
| AK295074.1 | ||||
| AK295760.1 | ||||
| AK314580.1 | ||||
| AL359945.1 | CAB95775.1 | |||
| BC030017.1 | AAH30017.1 | |||
| BC050409.1 | ||||
| CR936655.1 | CAI56793.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv574273 | Chr.17:4585589 - 5460332 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map