Assay Details
Target Gene Details
Entrez Gene ID: | 23452 |
Gene Name: | angiopoietin like 2 |
Gene Aliases: |
ARP2, HARP |
Location: |
Chr.9:127087349-127122803 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ANGPTL2 | NM_012098.2 | 2 | 738 | NP_036230.1 |
| XM_006717030.3 | 2 | 776 | XP_006717093.1 | |
| AF125175.1 | 1 | 259 | AAD55357.1 | |
| AK074726.1 | BAC11164.1 | |||
| AK307175.1 | ||||
| AK312876.1 | 2 | 657 | ||
| AY358274.1 | 2 | 757 | AAQ88641.1 | |
| BC012368.1 | 2 | 611 | AAH12368.1 |
Target Gene Details
Entrez Gene ID: | 9649 |
Gene Name: | Ral GEF with PH domain and SH3 binding motif 1 |
Gene Aliases: |
RALGEF2, RALGPS1A |
Location: |
Chr.9:126914774-127223166 on Build GRCh38 |
Assay Gene Location: | Within Intron 11 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv615342 | Chr.9:127091920 - 127178137 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map