Assay Details
Target Gene Details
Entrez Gene ID: | 55827 |
Gene Name: | DDB1 and CUL4 associated factor 6 |
Gene Aliases: |
1200006M05Rik, ARCAP, IQWD1, MSTP055, NRIP, PC326 |
Location: |
Chr.1:167936247-168075843 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DCAF6 | NM_001017977.2 | 1 | 111 | NP_001017977.1 |
| NM_001198956.1 | 1 | 111 | NP_001185885.1 | |
| NM_001198957.1 | 1 | 111 | NP_001185886.1 | |
| NM_018442.3 | 1 | 111 | NP_060912.2 | |
| XM_005245331.4 | 1 | 52 | XP_005245388.1 | |
| XM_005245332.4 | 1 | 52 | XP_005245389.1 | |
| XM_005245333.4 | 1 | 52 | XP_005245390.1 | |
| XM_017001779.1 | XP_016857268.1 | |||
| XM_017001780.1 | XP_016857269.1 | |||
| XM_017001781.1 | XP_016857270.1 | |||
| AK093970.1 | ||||
| AK226181.1 | ||||
| AK297847.1 | ||||
| BM822167.1 | ||||
| DA128283.1 | 1 | 111 |
Target Gene Details
Entrez Gene ID: | 25874 |
Gene Name: | mitochondrial pyruvate carrier 2 |
Gene Aliases: |
BRP44 |
Location: |
Chr.1:167916675-167937069 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MPC2 | NM_001143674.3 | NP_001137146.1 | ||
| NM_015415.3 | NP_056230.1 | |||
| XM_006711266.3 | 1 | 125 | XP_006711329.1 | |
| AL035304.1 | CAA22909.1 | |||
| AL554033.3 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv32660 | Chr.1:167825845 - 168220986 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map