Assay Details
Target Gene Details
Entrez Gene ID: | 100126784 |
Gene Name: | uncharacterized LOC100126784 |
Gene Aliases: |
- |
Location: |
Chr.11:19710934-19714672 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC100126784 | NR_015384.2 | 1 | 736 |
Target Gene Details
Entrez Gene ID: | 89797 |
Gene Name: | neuron navigator 2 |
Gene Aliases: |
HELAD1, POMFIL2, RAINB1, STEERIN2, UNC53H2 |
Location: |
Chr.11:19345200-20121601 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
More Information
Set Membership: |
|

Genomic Map