Assay Details
Target Gene Details
Entrez Gene ID: | 10488 |
Gene Name: | cAMP responsive element binding protein 3 |
Gene Aliases: |
LUMAN, LZIP, sLZIP |
Location: |
Chr.9:35732320-35737008 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CREB3 | NM_006368.4 | 1 | 50 | NP_006359.3 |
| AF211848.1 | AAG43528.1 | |||
| BG701230.1 | 1 | 55 |
Target Gene Details
Entrez Gene ID: | 7094 |
Gene Name: | talin 1 |
Gene Aliases: |
ILWEQ, TLN |
Location: |
Chr.9:35697337-35732395 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TLN1 | NM_006289.3 | NP_006280.3 | ||
| AB028950.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv25437 | Chr.9:35728113 - 35732419 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map