Assay Details
Target Gene Details
Entrez Gene ID: | 57082 |
Gene Name: | kinetochore scaffold 1 |
Gene Aliases: |
AF15Q14, CASC5, CT29, D40, MCPH4, PPP1R55, Spc7, hKNL-1, hSpc105 |
Location: |
Chr.15:40594012-40664342 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KNL1 | NM_144508.4 | 10 | 867 | NP_653091.3 |
| NM_170589.4 | 11 | 945 | NP_733468.3 | |
| XM_017022432.1 | 9 | 1070 | XP_016877921.1 | |
| AB022190.1 | 10 | 868 | BAC05691.1 | |
| AF173994.1 | 11 | 1174 | AAM45143.1 | |
| AF248041.1 | 10 | 851 | AAF97513.1 | |
| AF461041.1 | 10 | 851 | AAL67803.1 | |
| AK307605.1 | 10 | 859 | ||
| AK307996.1 | 10 | 857 | ||
| BC029373.1 | 10 | 844 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2760026 | Chr.15:40540346 - 40702913 on Build GRCh38 | Loss |
|
| nsv832984 | Chr.15:40502192 - 40706337 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs76371152] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map