Assay Details
Target Gene Details
Entrez Gene ID: | 9946 |
Gene Name: | crystallin zeta like 1 |
Gene Aliases: |
4P11, QOH-1 |
Location: |
Chr.21:33589341-33641854 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CRYZL1 | NM_145858.2 | NP_665857.2 | ||
| AF029689.1 | AAD22381.1 | |||
| AF098798.1 | AAD20219.1 | |||
| AK001293.1 | BAA91605.1 | |||
| AK057604.1 | 9 | 1310 | ||
| AK058071.1 | 4 | 338 | ||
| AK315707.1 | ||||
| BC033023.2 | AAH33023.1 | |||
| BI599245.1 | ||||
| BX647276.1 | 8 | 668 | ||
| BX648547.1 |
More Information
Set Membership: |
|

Genomic Map