Assay Details
Target Gene Details
Entrez Gene ID: | 50804 |
Gene Name: | myelin expression factor 2 |
Gene Aliases: |
HsT18564, MEF-2, MST156, MSTP156, myEF-2 |
Location: |
Chr.15:48134632-48178517 on Build GRCh38 |
Assay Gene Location: | Within Exon 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MYEF2 | NM_001301210.1 | 16 | 3599 | NP_001288139.1 |
| NM_016132.4 | 17 | 3671 | NP_057216.2 | |
| XM_005254422.4 | 16 | 3776 | XP_005254479.2 | |
| XM_005254424.4 | 15 | 3518 | XP_005254481.2 | |
| XM_005254425.4 | 16 | 3775 | XP_005254482.2 | |
| XM_006720553.3 | 16 | 3619 | XP_006720616.1 | |
| XM_011521657.2 | 13 | 3169 | XP_011519959.1 | |
| XM_017022285.1 | 15 | 3724 | XP_016877774.1 | |
| XM_017022286.1 | 15 | 3703 | XP_016877775.1 | |
| XM_017022287.1 | 14 | 3652 | XP_016877776.1 | |
| XM_017022288.1 | 15 | 3568 | XP_016877777.1 | |
| XM_017022289.1 | 15 | 3547 | XP_016877778.1 | |
| XM_017022290.1 | 14 | 3496 | XP_016877779.1 | |
| XM_017022291.1 | 14 | 3495 | XP_016877780.1 | |
| XM_017022292.1 | 13 | 3444 | XP_016877781.1 | |
| AB037762.1 | 17 | 3609 | ||
| BC152420.1 | 17 | 3609 | ||
| BC152451.1 | 17 | 3609 | ||
| BX647197.1 | 3 | 2784 |
Target Gene Details
Entrez Gene ID: | 283652 |
Gene Name: | solute carrier family 24 member 5 |
Gene Aliases: |
JSX, NCKX5, OCA6, SHEP4 |
Location: |
Chr.15:48120972-48142392 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 9 - Intron 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC24A5 | NM_205850.2 | NP_995322.1 | ||
| XM_017022079.1 | XP_016877568.1 | |||
| XM_017022080.1 | XP_016877569.1 | |||
| AF348468.1 | AAQ15116.1 | |||
| BC073944.1 | AAH73944.1 | |||
| BC113628.1 | ||||
| BC113630.1 | ||||
| BC143950.1 | ||||
| DQ665306.1 | ||||
| DQ665307.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832999 | Chr.15:47960097 - 48160208 on Build GRCh38 | Gain |
|
| nsv569338 | Chr.15:48104611 - 48160602 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map