Assay Details
Target Gene Details
Entrez Gene ID: | 100294362 |
Gene Name: | uncharacterized LOC100294362 |
Gene Aliases: |
- |
Location: |
Chr.17:80351830-80415168 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC100294362 | NR_029376.1 | |||
| AK125341.1 | BAC86138.1 |
Target Gene Details
Entrez Gene ID: | 57674 |
Gene Name: | ring finger protein 213 |
Gene Aliases: |
ALO17, C17orf27, KIAA1618, MYMY2, MYSTR, NET57 |
Location: |
Chr.17:80260820-80398786 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 47 - Exon 48 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RNF213 | NM_001256071.2 | NP_001243000.2 | ||
| XM_005257545.4 | XP_005257602.2 | |||
| XM_005257546.4 | XP_005257603.2 | |||
| XM_006721995.3 | XP_006722058.1 | |||
| XM_011525084.2 | XP_011523386.1 | |||
| XM_011525086.2 | XP_011523388.1 | |||
| XM_017024905.1 | XP_016880394.1 | |||
| AB046774.1 | BAB13380.1 | |||
| AB537889.1 | ||||
| AK074030.1 | BAB84856.1 | |||
| AK127358.1 | ||||
| AK127914.1 | BAC87188.1 | |||
| AL833201.1 | CAH56308.1 | |||
| BX640932.1 | CAE45967.1 | |||
| BX647946.1 | CAH56189.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs77506504] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map