Assay Details
Target Gene Details
Entrez Gene ID: | 1933 |
Gene Name: | eukaryotic translation elongation factor 1 beta 2 |
Gene Aliases: |
EEF1B, EEF1B1, EF1B |
Location: |
Chr.2:206159594-206162929 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EEF1B2 | NM_001037663.1 | NP_001032752.1 | ||
| NM_001959.3 | 1 | 295 | NP_001950.1 | |
| NM_021121.3 | NP_066944.1 | |||
| AK291910.1 | ||||
| AK310096.1 | ||||
| BC000211.1 | AAH00211.1 | |||
| BC067787.1 | AAH67787.1 | |||
| BM747256.1 | ||||
| D28350.1 | BAA05716.1 | |||
| S81522.1 | ||||
| X60489.1 | 1 | 144 | CAA43019.1 | |
| X60656.1 | CAA43063.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv821885 | Chr.2:206159334 - 206160501 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map