Assay Details
Target Gene Details
Entrez Gene ID: | 10201 |
Gene Name: | NME/NM23 nucleoside diphosphate kinase 6 |
Gene Aliases: |
IPIA-ALPHA, NDK 6, NM23-H6 |
Location: |
Chr.3:48289369-48302259 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 4 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834683 | Chr.3:48260014 - 48451855 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map