Assay Details
Target Gene Details
Entrez Gene ID: | 23529 |
Gene Name: | cardiotrophin-like cytokine factor 1 |
Gene Aliases: |
BSF-3, BSF3, CISS2, CLC, NNT-1, NNT1, NR6 |
Location: |
Chr.11:67364168-67374177 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CLCF1 | NM_001166212.1 | 3 | 721 | NP_001159684.1 |
| NM_013246.2 | 3 | 802 | NP_037378.1 | |
| AF172854.1 | 3 | 651 | AAD54284.1 | |
| AF176911.1 | 3 | 695 | AAF00991.1 | |
| AK298052.1 | 3 | 721 | ||
| AY049779.1 | 3 | 608 | AAL15436.1 | |
| BC012939.1 | 3 | 678 | AAH12939.1 | |
| BC066229.1 | 3 | 610 | AAH66229.1 | |
| BC066230.1 | 3 | 610 | AAH66230.1 | |
| BC066231.1 | 3 | 610 | AAH66231.1 |
Target Gene Details
Entrez Gene ID: | 100130987 |
Gene Name: | uncharacterized LOC100130987 |
Gene Aliases: |
- |
Location: |
Chr.11:67317839-67391687 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC100130987 | NR_024469.1 | |||
| AK096155.1 | ||||
| DA400240.1 | ||||
| DB058546.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv468603 | Chr.11:67265605 - 67397024 on Build GRCh38 | Loss |
|
| dgv52n111 | Chr.11:67362651 - 67521123 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map